The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286250.1 | NP_001273179.1 | 2 - 3 | 494 | 64 |
NM_001286251.1 | NP_001273180.1 | 2 - 3 | 494 | 64 | |
NM_001286252.1 | NP_001273181.1 | 2 - 3 | 494 | 64 | |
NM_001323252.1 | NP_001310181.1 | 3 - 4 | 541 | 64 | |
NM_001323253.1 | NP_001310182.1 | 2 - 3 | 494 | 64 | |
NM_001323254.1 | NP_001310183.1 | 2 - 3 | 494 | 64 | |
NM_001323255.1 | NP_001310184.1 | 2 - 3 | 494 | 64 | |
NM_001323256.1 | NP_001310185.1 | 2 - 3 | 494 | 64 | |
NM_001323257.1 | NP_001310186.1 | 2 - 3 | 494 | 64 | |
NM_001323258.1 | NP_001310187.1 | 3 - 4 | 2238 | 64 | |
NM_001323259.1 | NP_001310188.1 | 2 - 3 | 494 | 64 | |
NM_001323260.1 | NP_001310189.1 | 2 - 3 | 494 | 64 | |
NM_052904.3 | NP_443136.2 | 2 - 3 | 494 | 64 | |
NR_104421.1 | 2 - 3 | 494 | 64 | ||
NR_136580.1 | 2 - 3 | 494 | 64 | ||
GenBank mRNA | AF351610.1 | - | 2 - 3 | 322 | 64 |
AK055292.1 | - | 2 - 3 | 380 | 64 | |
AK295479.1 | - | 2 - 3 | 319 | 64 | |
AK297240.1 | - | 2 - 3 | 238 | 64 | |
BC031830.1 | - | 1 - 2 | 75 | 64 | |
BC071627.1 | - | 2 - 3 | 369 | 64 | |
DA296671.1 | - | 2 - 3 | 386 | 64 | |
DA332224.1 | - | 3 - 4 | 266 | 64 | |
DC314842.1 | - | 2 - 3 | 489 | 64 |