The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286250.1 | NP_001273179.1 | 9 - 10 | 2058 | 113 |
NM_001286251.1 | NP_001273180.1 | 8 - 9 | 1959 | 113 | |
NM_001286254.2 | NP_001273183.1 | 8 - 9 | 1124 | 113 | |
NM_001323252.1 | NP_001310181.1 | 11 - 12 | 2213 | 113 | |
NM_001323257.1 | NP_001310186.1 | 9 - 10 | 1950 | 113 | |
NM_001323260.1 | NP_001310189.1 | 10 - 11 | 2162 | 113 | |
NM_001323261.1 | NP_001310190.1 | 8 - 9 | 1676 | 113 | |
NM_001323263.1 | NP_001310192.1 | 8 - 9 | 1680 | 113 | |
NM_001323264.1 | NP_001310193.1 | 7 - 8 | 1464 | 113 | |
NM_052904.3 | NP_443136.2 | 10 - 11 | 2166 | 113 | |
NR_104421.1 | 8 - 9 | 1223 | 113 | ||
NR_136579.1 | 7 - 8 | 953 | 113 | ||
NR_136580.1 | 9 - 10 | 1439 | 113 | ||
XM_005266813.4 | XP_005266870.1 | 8 - 9 | 1571 | 113 | |
XM_017010228.1 | XP_016865717.1 | 7 - 8 | 1355 | 113 | |
XM_017010229.1 | XP_016865718.1 | 8 - 9 | 1528 | 113 | |
GenBank mRNA | AB067487.1 | - | 8 - 9 | 1580 | 113 |
AF351610.1 | - | 8 - 9 | 1052 | 113 | |
AK055292.1 | - | 10 - 11 | 2052 | 113 | |
AK295479.1 | - | 9 - 10 | 1883 | 113 | |
AK297240.1 | - | 8 - 9 | 1703 | 113 | |
BC031830.1 | - | 7 - 8 | 804 | 113 | |
BC047491.1 | - | 8 - 9 | 1004 | 113 |