The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052925.3 | NP_443157.1 | 11 - 12 | 1933 | 124 |
XM_005278248.4 | XP_005278305.1 | 11 - 12 | 1924 | 124 | |
XM_005278249.4 | XP_005278306.1 | 11 - 12 | 2376 | 124 | |
XM_005278250.4 | XP_005278307.1 | 10 - 11 | 1795 | 124 | |
XM_005278252.4 | XP_005278309.1 | 10 - 11 | 2265 | 124 | |
XM_006722996.3 | XP_006723059.1 | 10 - 11 | 1800 | 124 | |
XM_006722997.3 | XP_006723060.1 | 9 - 10 | 1689 | 124 | |
XM_011526413.2 | XP_011524715.1 | 11 - 12 | 1863 | 124 | |
XM_011526414.2 | XP_011524716.1 | 10 - 11 | 1813 | 124 | |
XM_011526415.2 | XP_011524717.1 | 11 - 12 | 1924 | 124 | |
GenBank mRNA | AB067519.1 | - | - | 919 | 124 |
BC028048.1 | - | 11 - 12 | 1884 | 124 |