The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_058238.2 | NP_478679.1 | 2 - 3 | 675 | 79 |
XM_011530366.1 | XP_011528668.1 | 2 - 3 | 440 | 79 | |
GenBank mRNA | AB062766.1 | - | 2 - 3 | 306 | 79 |
AF416743.1 | - | 2 - 3 | 675 | 79 | |
AK123019.1 | - | 2 - 3 | 270 | 79 | |
AK289526.1 | - | 2 - 3 | 440 | 79 | |
BC034923.1 | - | 2 - 3 | 398 | 79 | |
BM047487.1 | - | 2 - 3 | 397 | 78 |