The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138409.2 | NP_612418.2 | 1 - 2 | 178 | 72 |
XM_017010221.1 | XP_016865710.1 | 1 - 2 | 479 | 72 | |
GenBank mRNA | AK090775.1 | - | 1 - 2 | 178 | 72 |
AK292736.1 | - | 1 - 2 | 109 | 72 | |
AK307078.1 | - | 1 - 2 | 107 | 72 | |
BC010003.2 | - | 1 - 2 | 123 | 72 |