The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138409.2 | NP_612418.2 | 3 - 4 | 414 | 92 |
XM_011535400.2 | XP_011533702.1 | 3 - 4 | 340 | 92 | |
XM_017010219.1 | XP_016865708.1 | 4 - 5 | 712 | 92 | |
XM_017010220.1 | XP_016865709.1 | 3 - 4 | 981 | 92 | |
GenBank mRNA | AK090775.1 | - | 3 - 4 | 414 | 92 |
AK292736.1 | - | 3 - 4 | 345 | 92 | |
BC010003.2 | - | 3 - 4 | 359 | 92 | |
BC039855.1 | - | 3 - 4 | 277 | 92 |