The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138433.3 | NP_612442.2 | 1 - 1 | 1480 | 68 |
GenBank mRNA | BC009980.2 | - | 1 - 1 | 1480 | 68 |