The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138434.2 | NP_612443.1 | 1 - 1 | 918 | 126 |
GenBank mRNA | BC011406.1 | - | 1 - 1 | 455 | 126 |