The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006277.2 | NP_006268.2 | 7 - 8 | 909 | 70 |
NM_019595.3 | NP_062541.3 | 7 - 8 | 909 | 70 | |
NM_147152.2 | NP_671494.2 | 7 - 8 | 909 | 70 | |
GenBank mRNA | AB033082.1 | - | 7 - 8 | 816 | 70 |
AF182198.1 | - | 7 - 8 | 888 | 70 | |
AF182199.1 | - | 7 - 8 | 888 | 70 | |
AF248540.1 | - | 6 - 7 | 690 | 70 | |
BC020921.2 | - | 6 - 7 | 686 | 70 | |
BC146779.1 | - | 7 - 8 | 816 | 70 |