The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152334.2 | NP_689547.2 | 13 - 14 | 1845 | 93 |
XM_006720386.3 | XP_006720449.1 | 13 - 14 | 2007 | 93 | |
XM_017021912.1 | XP_016877401.1 | 13 - 14 | 2007 | 93 | |
XM_017021913.1 | XP_016877402.1 | 10 - 11 | 1295 | 93 | |
GenBank mRNA | AK056653.1 | - | 11 - 12 | 1550 | 93 |
AK057734.1 | - | 7 - 8 | 848 | 93 | |
AL833188.1 | - | 13 - 14 | 1511 | 93 | |
BC069346.1 | - | 6 - 7 | 783 | 93 | |
BC069811.1 | - | 6 - 7 | 783 | 93 | |
BC074887.2 | - | 5 - 6 | 709 | 93 | |
BC132671.1 | - | 13 - 14 | 1813 | 93 | |
BC136315.1 | - | 13 - 14 | 1813 | 93 | |
BC143943.1 | - | 13 - 14 | 1813 | 93 |