The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114106.2 | NP_001107578.1 | 1 - 2 | 134 | 106 |
NM_001258340.1 | NP_001245269.1 | 1 - 2 | 134 | 106 | |
NM_001258341.1 | NP_001245270.1 | 1 - 2 | 134 | 106 | |
XM_005270440.2 | XP_005270497.1 | 1 - 2 | 125 | 106 | |
XM_005270442.1 | XP_005270499.1 | 1 - 2 | 125 | 106 | |
XM_011540635.2 | XP_011538937.1 | 1 - 2 | 125 | 106 | |
XM_011540636.2 | XP_011538938.1 | 1 - 2 | 125 | 106 | |
XM_017000264.1 | XP_016855753.1 | 1 - 2 | 125 | 106 | |
XM_017000265.1 | XP_016855754.1 | 1 - 2 | 125 | 106 | |
XM_017000266.1 | XP_016855755.1 | 1 - 2 | 125 | 106 | |
GenBank mRNA | AK316123.1 | - | 1 - 2 | 75 | 106 |
AL530322.3 | - | 1 - 2 | 126 | 106 | |
BC033858.1 | - | 1 - 2 | 97 | 106 | |
DC311042.1 | - | 1 - 2 | 131 | 106 |