The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114106.2 | NP_001107578.1 | 3 - 4 | 393 | 80 |
NM_001258340.1 | NP_001245269.1 | 3 - 4 | 393 | 80 | |
NM_001258342.1 | NP_001245271.1 | 2 - 3 | 285 | 80 | |
NM_152369.4 | NP_689582.2 | 2 - 3 | 328 | 80 | |
XM_005270441.2 | XP_005270498.1 | 3 - 4 | 330 | 80 | |
XM_005270442.1 | XP_005270499.1 | 3 - 4 | 384 | 80 | |
XM_011540635.2 | XP_011538937.1 | 3 - 4 | 384 | 80 | |
XM_017000262.1 | XP_016855751.1 | 2 - 3 | 270 | 80 | |
XM_017000263.1 | XP_016855752.1 | 3 - 4 | 330 | 80 | |
XM_017000264.1 | XP_016855753.1 | 3 - 4 | 384 | 80 | |
XM_017000266.1 | XP_016855755.1 | 3 - 4 | 384 | 80 | |
GenBank mRNA | AK303906.1 | - | 2 - 3 | 264 | 80 |
AK313574.1 | - | 3 - 4 | 281 | 80 | |
AL530322.3 | - | 3 - 4 | 385 | 80 | |
BC033858.1 | - | 3 - 4 | 356 | 80 | |
BC053877.1 | - | 2 - 3 | 251 | 80 | |
DA538278.1 | - | 2 - 3 | 328 | 80 |