Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SLC44A3
- Entrez Gene ID
-
126969
- Gene Name
- solute carrier family 44 member 3
- Gene Aliases
-
CTL3
- Chromosome Location
-
Chr.1: 94820342 - 94895247 on Build GRCh38
View Full Product Details