The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152380.2 | NP_689593.2 | 7 - 8 | 1021 | 72 |
XM_005271162.1 | XP_005271219.1 | 7 - 8 | 1477 | 72 | |
GenBank mRNA | AK096396.1 | - | 3 - 4 | 308 | 72 |
AK127536.1 | - | 7 - 8 | 1021 | 72 | |
BC122553.1 | - | 7 - 8 | 839 | 72 |