The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039693.2 | NP_001034782.1 | 5 - 6 | 755 | 129 |
NR_004862.1 | 5 - 6 | 736 | 129 | ||
NR_109907.1 | 4 - 5 | 600 | 129 | ||
XM_017003310.1 | XP_016858799.1 | 3 - 4 | 383 | 129 | |
GenBank mRNA | AK095272.1 | - | 4 - 5 | 364 | 129 |
AK298977.1 | - | 5 - 6 | 499 | 129 | |
BC132835.1 | - | 5 - 6 | 519 | 129 | |
BC136837.1 | - | 5 - 6 | 519 | 129 | |
BC144420.1 | - | 5 - 6 | 500 | 129 | |
BX482572.1 | - | 5 - 6 | 509 | 129 | |
BX647677.1 | - | 5 - 6 | 737 | 129 | |
CN261486.1 | - | 5 - 6 | 468 | 129 |