The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026849.1 | 1 - 1 | 854 | 58 | |
GenBank mRNA | AK292743.1 | - | 1 - 1 | 1431 | 58 |
BC010538.1 | - | 1 - 1 | 854 | 58 |