Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDSUB1
Entrez Gene ID
151525
Gene Name
WD repeat, sterile alpha motif and U-box domain containing 1
Gene Aliases
UBOX6, WDSAM1
Chromosome Location
Chr.2: 159235793 - 159286799 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128212.2 NP_001121684.1 7 - 8 1080 105
NM_001128213.1 NP_001121685.1 7 - 8 1096 105
NM_152528.2 NP_689741.2 7 - 8 988 105
XM_005246355.2 XP_005246412.2 7 - 8 1074 105
XM_006712341.3 XP_006712404.1 6 - 7 568 105
XM_011510753.2 XP_011509055.1 7 - 8 1040 105
XM_011510754.2 XP_011509056.1 7 - 8 774 105
XM_017003481.1 XP_016858970.1 7 - 8 1074 105
GenBank mRNA AK093494.1 - 7 - 8 988 105
AK129983.1 - 7 - 8 1096 105
AL040518.1 - 6 - 7 464 105

View Full Product Details