The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287499.1 | NP_001274428.1 | 11 - 12 | 1030 | 61 |
NM_001287500.1 | NP_001274429.1 | 10 - 11 | 982 | 61 | |
NM_001287501.1 | NP_001274430.1 | 9 - 10 | 936 | 61 | |
NM_001287502.1 | NP_001274431.1 | 9 - 10 | 936 | 61 | |
NM_152558.4 | NP_689771.3 | 11 - 12 | 1030 | 61 | |
XM_006715676.2 | XP_006715739.1 | 9 - 10 | 784 | 61 | |
XM_011515242.1 | XP_011513544.1 | 10 - 11 | 889 | 61 | |
XM_011515243.1 | XP_011513545.1 | 9 - 10 | 911 | 61 | |
XM_011515244.2 | XP_011513546.1 | 4 - 5 | 325 | 61 | |
XM_017011902.1 | XP_016867391.1 | 11 - 12 | 1092 | 61 | |
XM_017011903.1 | XP_016867392.1 | 11 - 12 | 970 | 61 | |
GenBank mRNA | AB028946.2 | - | 9 - 10 | 733 | 61 |
AK092404.1 | - | 11 - 12 | 924 | 61 | |
AK293372.1 | - | 10 - 11 | 805 | 61 | |
AK302053.1 | - | 9 - 10 | 884 | 61 | |
AK309646.1 | - | 11 - 12 | 954 | 61 | |
AL136792.1 | - | 10 - 11 | 711 | 61 | |
BC043150.1 | - | 9 - 10 | 856 | 61 | |
BC061518.1 | - | 10 - 11 | 866 | 61 | |
BC071858.1 | - | 11 - 12 | 1004 | 61 | |
KJ534872.1 | - | 11 - 12 | 820 | 61 | |
KJ535081.1 | - | 9 - 10 | 726 | 61 |