The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287499.1 | NP_001274428.1 | 16 - 17 | 1657 | 84 |
NM_001287500.1 | NP_001274429.1 | 15 - 16 | 1609 | 84 | |
NM_001287501.1 | NP_001274430.1 | 14 - 15 | 1563 | 84 | |
NM_001287502.1 | NP_001274431.1 | 14 - 15 | 1563 | 84 | |
NM_152558.4 | NP_689771.3 | 16 - 17 | 1657 | 84 | |
XM_006715676.2 | XP_006715739.1 | 14 - 15 | 1411 | 84 | |
XM_011515242.1 | XP_011513544.1 | 15 - 16 | 1516 | 84 | |
XM_011515243.1 | XP_011513545.1 | 14 - 15 | 1538 | 84 | |
XM_011515244.2 | XP_011513546.1 | 9 - 10 | 952 | 84 | |
XM_017011902.1 | XP_016867391.1 | 16 - 17 | 1719 | 84 | |
XM_017011903.1 | XP_016867392.1 | 16 - 17 | 1597 | 84 | |
GenBank mRNA | AB028946.2 | - | 14 - 15 | 1360 | 84 |
AK092404.1 | - | 16 - 17 | 1551 | 84 | |
AK293372.1 | - | 15 - 16 | 1432 | 84 | |
AK302053.1 | - | 14 - 15 | 1511 | 84 | |
AL136792.1 | - | 15 - 16 | 1338 | 84 | |
BC043150.1 | - | 14 - 15 | 1483 | 84 | |
BC071858.1 | - | 16 - 17 | 1631 | 84 | |
KJ534872.1 | - | 16 - 17 | 1447 | 84 | |
KJ535081.1 | - | 14 - 15 | 1353 | 84 |