The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287499.1 | NP_001274428.1 | 19 - 20 | 1954 | 140 |
NM_001287500.1 | NP_001274429.1 | 18 - 19 | 1906 | 140 | |
NM_001287501.1 | NP_001274430.1 | 17 - 18 | 1860 | 140 | |
NM_001287502.1 | NP_001274431.1 | 17 - 18 | 1860 | 140 | |
NM_152558.4 | NP_689771.3 | 19 - 20 | 1954 | 140 | |
XM_006715676.2 | XP_006715739.1 | 17 - 18 | 1708 | 140 | |
XM_011515242.1 | XP_011513544.1 | 18 - 19 | 1813 | 140 | |
XM_011515243.1 | XP_011513545.1 | 17 - 18 | 1835 | 140 | |
XM_011515244.2 | XP_011513546.1 | 12 - 13 | 1249 | 140 | |
XM_017011902.1 | XP_016867391.1 | 19 - 20 | 2016 | 140 | |
XM_017011903.1 | XP_016867392.1 | 19 - 20 | 1894 | 140 | |
GenBank mRNA | AB028946.2 | - | 17 - 18 | 1657 | 140 |
AK092404.1 | - | 19 - 20 | 1848 | 140 | |
AK293372.1 | - | 18 - 19 | 1729 | 140 | |
AK302053.1 | - | 17 - 18 | 1808 | 140 | |
AL136792.1 | - | 18 - 19 | 1635 | 140 | |
BC043150.1 | - | 17 - 18 | 1780 | 140 | |
BC071858.1 | - | 19 - 20 | 140 | ||
DA246430.1 | - | 2 - 3 | 132 | 140 | |
KJ534872.1 | - | 19 - 20 | 1744 | 140 | |
KJ535081.1 | - | 17 - 18 | 1650 | 140 |