The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152620.2 | NP_689833.1 | 1 - 2 | 114 | 118 |
GenBank mRNA | AK093201.1 | - | 1 - 2 | 115 | 118 |