The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195811.1 | NP_001182740.1 | 4 - 5 | 545 | 88 |
NM_001195812.1 | NP_001182741.1 | 3 - 4 | 383 | 88 | |
NM_001320272.1 | NP_001307201.1 | 5 - 6 | 839 | 88 | |
NM_152666.2 | NP_689879.2 | 5 - 6 | 832 | 88 | |
XM_006711752.2 | XP_006711815.1 | 4 - 5 | 999 | 88 | |
XM_011544115.2 | XP_011542417.1 | 6 - 7 | 907 | 88 | |
XM_011544116.2 | XP_011542418.1 | 6 - 7 | 735 | 88 | |
XM_017000567.1 | XP_016856056.1 | 4 - 5 | 1012 | 88 | |
XM_017000568.1 | XP_016856057.1 | 4 - 5 | 533 | 88 | |
GenBank mRNA | AK091691.1 | - | 3 - 4 | 357 | 88 |
AK098092.1 | - | 5 - 6 | 839 | 88 | |
AK295421.1 | - | 4 - 5 | 526 | 88 | |
AK307857.1 | - | 4 - 5 | 524 | 88 | |
AY391836.1 | - | 3 - 4 | 395 | 88 | |
AY461578.1 | - | 4 - 5 | 545 | 88 | |
BC101373.1 | - | 4 - 5 | 472 | 88 | |
BC101374.1 | - | 4 - 5 | 472 | 88 | |
BC101375.1 | - | 4 - 5 | 472 | 88 |