Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPERT
Entrez Gene ID
220082
Gene Name
spermatid associated
Gene Aliases
CBY2, NURIT
Chromosome Location
Chr.13: 45701030 - 45714559 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152719.2 NP_689932.1 2 - 3 241 146
GenBank mRNA AK093129.1 - 2 - 3 234 146
HY007057.1 - 2 - 3 233 146
HY017873.1 - 2 - 3 243 146

View Full Product Details