Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APCDD1
Entrez Gene ID
147495
Gene Name
APC down-regulated 1
Gene Aliases
B7323, DRAPC1, FP7019, HHS, HTS, HYPT1
Chromosome Location
Chr.18: 10454628 - 10488701 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153000.4 NP_694545.1 3 - 4 1127 73
GenBank mRNA AB056722.1 - 3 - 4 1126 73
AB104887.1 - 3 - 4 1126 73
AF461902.1 - 2 - 3 723 73
AK300743.1 - 4 - 5 929 73
AK303076.1 - 2 - 3 719 73
BC053324.1 - 4 - 5 1098 73

View Full Product Details