The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AF520800.1 | - | 4 - 5 | 344 | 85 |
AF520803.1 | - | 2 - 3 | 216 | 85 | |
AF520804.1 | - | 2 - 3 | 202 | 85 | |
AF520805.1 | - | 2 - 3 | 216 | 85 | |
AF520806.1 | - | 2 - 3 | 224 | 85 | |
AF520807.1 | - | 3 - 4 | 274 | 85 | |
AF520808.1 | - | 3 - 4 | 304 | 85 | |
AF548113.1 | - | 4 - 5 | 490 | 85 | |
AF548118.1 | - | - | 177 | 85 | |
AF548122.1 | - | 1 - 2 | 177 | 85 | |
AF548123.1 | - | 1 - 2 | 177 | 85 | |
AY309094.1 | - | 5 - 6 | 425 | 85 | |
BC113541.1 | - | 3 - 4 | 283 | 85 | |
BC113543.1 | - | 3 - 4 | 283 | 85 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017011612.1 | XP_016867101.1 | 5 - 6 | 641 | 85 |