The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286434.1 | NP_001273363.1 | 4 - 5 | 493 | 71 |
NM_153356.2 | NP_699187.1 | 5 - 6 | 601 | 71 | |
XM_006720409.3 | XP_006720472.1 | 5 - 6 | 631 | 71 | |
XM_006720410.3 | XP_006720473.1 | 4 - 5 | 523 | 71 | |
XM_006720411.3 | XP_006720474.1 | 3 - 4 | 419 | 71 | |
XM_011521281.2 | XP_011519583.1 | 5 - 6 | 630 | 71 | |
XM_011521282.2 | XP_011519584.1 | 5 - 6 | 630 | 71 | |
XM_011521283.2 | XP_011519585.1 | 5 - 6 | 630 | 71 | |
XM_011521284.2 | XP_011519586.1 | 4 - 5 | 410 | 71 | |
GenBank mRNA | AB449907.1 | - | 4 - 5 | 369 | 71 |
BC033516.1 | - | 5 - 6 | 560 | 71 |