The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014620.5 | NP_055435.2 | 3 - 4 | 1128 | 85 |
NM_153633.2 | NP_705897.1 | 1 - 2 | 488 | 85 | |
GenBank mRNA | X07495.1 | - | 3 - 4 | 1049 | 85 |