Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HTRA4
Entrez Gene ID
203100
Gene Name
HtrA serine peptidase 4
Gene Aliases
-
Chromosome Location
Chr.8: 38974149 - 38988662 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153692.3 NP_710159.1 7 - 8 1281 120
GenBank mRNA AK075205.1 - 7 - 8 1252 120
AK300197.1 - 7 - 8 1160 120
BC057765.1 - 7 - 8 1213 120

View Full Product Details