Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FADD
Entrez Gene ID
8772
Gene Name
Fas associated via death domain
Gene Aliases
GIG3, MORT1
Chromosome Location
Chr.11: 70203163 - 70207402 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003824.3 NP_003815.1 1 - 2 582 112
GenBank mRNA AK291005.1 - 1 - 2 443 112
AY423721.1 - 1 - 2 112
BC000334.2 - 1 - 2 430 112
BC025733.1 - 1 - 2 406 112
BT006927.1 - 1 - 2 285 112
CR456738.1 - 1 - 2 285 112
U24231.1 - 1 - 2 414 112
X84709.1 - 1 - 2 429 112

View Full Product Details