Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNRNP200
Entrez Gene ID
23020
Gene Name
small nuclear ribonucleoprotein U5 subunit 200
Gene Aliases
ASCC3L1, BRR2, HELIC2, RP33, U5-200KD
Chromosome Location
Chr.2: 96274336 - 96321893 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014014.4 NP_054733.2 38 - 39 5618 65
XM_017003603.1 XP_016859092.1 40 - 41 6448 65
GenBank mRNA AB018331.2 - 36 - 37 5156 65
AF085356.1 - 8 - 9 1460 65
AF119874.1 - 2 - 3 191 65
AK023840.1 - 32 - 33 4629 65
AK024391.1 - 32 - 33 4699 65
AK026662.1 - 6 - 7 740 65
AK303465.1 - 9 - 10 1455 65
AK303663.1 - 9 - 10 1307 65
AL831994.1 - 29 - 30 4334 65
AY572488.1 - 38 - 39 5564 65
BC001417.2 - 4 - 5 562 65
BC007577.1 - 20 - 21 3002 65
BC065924.1 - 31 - 32 4541 65
BC112891.1 - 32 - 33 4739 65
BC131784.1 - 8 - 9 1150 65

View Full Product Details