The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308142.1 | NP_001295071.1 | 17 - 17 | 7804 | 113 |
NM_014048.4 | NP_054767.3 | 17 - 17 | 7655 | 113 | |
XM_005255452.3 | XP_005255509.1 | 18 - 18 | 7959 | 113 | |
XM_005255453.4 | XP_005255510.1 | 17 - 17 | 8443 | 113 | |
XM_005255455.3 | XP_005255512.1 | 15 - 15 | 7789 | 113 | |
XM_006720908.3 | XP_006720971.1 | 18 - 18 | 8605 | 113 | |
XM_006720909.3 | XP_006720972.1 | 18 - 18 | 7934 | 113 | |
XM_006720912.3 | XP_006720975.1 | 15 - 15 | 9744 | 113 | |
XM_006720913.2 | XP_006720976.1 | 17 - 17 | 7821 | 113 | |
XM_006720914.2 | XP_006720977.1 | 18 - 18 | 7809 | 113 | |
XM_011522568.2 | XP_011520870.1 | 17 - 17 | 7774 | 113 | |
XM_011522569.2 | XP_011520871.1 | 15 - 15 | 7571 | 113 | |
XM_017023501.1 | XP_016878990.1 | 17 - 17 | 7770 | 113 | |
XM_017023502.1 | XP_016878991.1 | 17 - 17 | 7827 | 113 | |
XM_017023503.1 | XP_016878992.1 | 16 - 16 | 10972 | 113 | |
XM_017023504.1 | XP_016878993.1 | 15 - 15 | 7639 | 113 | |
XM_017023505.1 | XP_016878994.1 | 15 - 15 | 9594 | 113 | |
GenBank mRNA | AB033069.2 | - | 9 - 9 | 6811 | 113 |
AF078853.1 | - | 1 - 1 | 115 | ||
AK056385.1 | - | 1 - 1 | 2137 | 113 | |
AK057216.1 | - | 1 - 1 | 1171 | 113 | |
BC006977.1 | - | 1 - 1 | 356 | 113 | |
BC016293.1 | - | 1 - 1 | 579 | 113 | |
BC030510.1 | - | 1 - 1 | 449 | 113 | |
BC050541.1 | - | 1 - 1 | 279 | 113 | |
BC107868.1 | - | 1 - 1 | 584 | 113 | |
BX641041.1 | - | 5 - 5 | 6892 | 113 | |
CR749797.1 | - | 6 - 6 | 6350 | 113 |