The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284423.1 | NP_001271352.1 | 9 - 10 | 1118 | 87 |
NM_001284424.1 | NP_001271353.1 | 9 - 10 | 1307 | 87 | |
NM_001284425.1 | NP_001271354.1 | 8 - 9 | 1170 | 87 | |
NM_001284426.1 | NP_001271355.1 | 7 - 8 | 928 | 87 | |
NM_015460.3 | NP_056275.2 | 9 - 10 | 1307 | 87 | |
NR_104316.1 | 9 - 10 | 1307 | 87 | ||
XM_011533575.1 | XP_011531877.1 | 9 - 10 | 1279 | 87 | |
GenBank mRNA | AB083783.1 | - | 8 - 9 | 1020 | 87 |
AF396687.1 | - | 9 - 10 | 1156 | 87 | |
AK125334.1 | - | 7 - 8 | 794 | 87 | |
AK126013.1 | - | 9 - 10 | 1261 | 87 | |
AK294714.1 | - | 7 - 8 | 918 | 87 | |
AK312412.1 | - | 9 - 10 | 1117 | 87 | |
AK316068.1 | - | 7 - 8 | 928 | 87 | |
BC092512.1 | - | 9 - 10 | 1306 | 87 | |
BC109311.1 | - | 8 - 9 | 998 | 87 | |
BC109312.1 | - | 9 - 10 | 1135 | 87 |