The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284423.1 | NP_001271352.1 | 15 - 16 | 2525 | 107 |
NM_001284424.1 | NP_001271353.1 | 14 - 15 | 2519 | 107 | |
NM_001284425.1 | NP_001271354.1 | 14 - 15 | 2577 | 107 | |
NM_001284426.1 | NP_001271355.1 | 13 - 14 | 2335 | 107 | |
NM_015460.3 | NP_056275.2 | 15 - 16 | 2714 | 107 | |
NR_104316.1 | 15 - 16 | 2677 | 107 | ||
XM_011533575.1 | XP_011531877.1 | 15 - 16 | 2686 | 107 | |
GenBank mRNA | AB083783.1 | - | 14 - 15 | 2427 | 107 |
AF396687.1 | - | 15 - 16 | 2563 | 107 | |
AK125334.1 | - | 13 - 14 | 2201 | 107 | |
AK294714.1 | - | 13 - 14 | 2325 | 107 | |
AK312412.1 | - | 15 - 16 | 2524 | 107 | |
AK316068.1 | - | 13 - 14 | 2335 | 107 | |
AL050090.1 | - | 3 - 4 | 333 | 107 | |
BC092512.1 | - | 15 - 16 | 2713 | 107 | |
BC109311.1 | - | 14 - 15 | 2405 | 107 | |
BC109312.1 | - | 15 - 16 | 2505 | 107 |