Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTMR12
Entrez Gene ID
54545
Gene Name
myotubularin related protein 12
Gene Aliases
3-PAP, PIP3AP
Chromosome Location
Chr.5: 32227007 - 32313008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040446.2 NP_001035536.1 1 - 2 255 91
NM_001294343.1 NP_001281272.1 1 - 2 255 91
NM_001294344.1 NP_001281273.1 1 - 2 255 91
XM_005248313.3 XP_005248370.1 1 - 2 219 91
XM_011514057.1 XP_011512359.1 1 - 2 219 91
XM_011514058.2 XP_011512360.1 1 - 2 219 91
XM_011514059.1 XP_011512361.1 1 - 2 219 91
XM_011514060.1 XP_011512362.1 1 - 2 219 91
XM_017009589.1 XP_016865078.1 1 - 2 219 91
XM_017009593.1 XP_016865082.1 1 - 2 219 91
XM_017009594.1 XP_016865083.1 1 - 2 235 91
XM_017009596.1 XP_016865085.1 1 - 2 171 91
GenBank mRNA AB051469.1 - 1 - 2 188 91
AK000483.1 - 1 - 2 216 91
AY028703.1 - 1 - 2 192 91
BC057393.1 - 1 - 2 185 91
DB462159.1 - 1 - 2 258 91
DB483084.1 - 1 - 2 255 91

View Full Product Details