The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142643.2 | NP_001136115.1 | 10 - 11 | 1259 | 79 |
NM_020753.4 | NP_065804.2 | 11 - 12 | 1776 | 79 | |
GenBank mRNA | AB032965.1 | - | 7 - 8 | 955 | 79 |
AF451976.1 | - | 11 - 12 | 1629 | 79 | |
AK300354.1 | - | 10 - 11 | 1206 | 79 | |
AK315936.1 | - | 10 - 11 | 1260 | 79 | |
BC066643.1 | - | 11 - 12 | 1739 | 79 |