The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142643.2 | NP_001136115.1 | 15 - 16 | 1757 | 57 |
NM_020753.4 | NP_065804.2 | 16 - 17 | 2274 | 57 | |
GenBank mRNA | AB032965.1 | - | 12 - 13 | 1453 | 57 |
AF451976.1 | - | 16 - 17 | 2127 | 57 | |
AK300354.1 | - | 15 - 16 | 1704 | 57 | |
AK315936.1 | - | 15 - 16 | 1758 | 57 | |
BC066643.1 | - | 16 - 17 | 2237 | 57 |