The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017553.2 | NP_060023.1 | 36 - 36 | 5843 | 70 |
NR_104038.1 | 35 - 35 | 5715 | 70 | ||
XM_011521686.2 | XP_011519988.1 | 22 - 22 | 3654 | 70 | |
GenBank mRNA | AK002176.1 | - | 8 - 8 | 2006 | 70 |
AK024777.1 | - | 1 - 1 | 649 | 70 | |
AL137280.1 | - | 5 - 5 | 1622 | 70 | |
BC046115.2 | - | 1 - 1 | 647 | 70 | |
BC057299.1 | - | 1 - 1 | 747 | 70 | |
BC105993.1 | - | 1 - 1 | 656 | 70 |