Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHBDD1
Entrez Gene ID
84236
Gene Name
rhomboid domain containing 1
Gene Aliases
RRP4
Chromosome Location
Chr.2: 226800164 - 226999210 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167608.1 NP_001161080.1 9 - 9 2007 93
NM_032276.3 NP_115652.2 7 - 7 1706 93
XM_017005083.1 XP_016860572.1 9 - 9 2001 93
XM_017005084.1 XP_016860573.1 9 - 9 3076 93
XM_017005085.1 XP_016860574.1 8 - 8 3062 93
XM_017005086.1 XP_016860575.1 8 - 8 2994 93
XM_017005087.1 XP_016860576.1 7 - 7 2843 93
XM_017005088.1 XP_016860577.1 8 - 8 1782 93
XM_017005089.1 XP_016860578.1 9 - 9 2021 93
XM_017005090.1 XP_016860579.1 8 - 8 2857 93
XM_017005091.1 XP_016860580.1 8 - 8 1687 93
XM_017005092.1 XP_016860581.1 7 - 7 2775 93
GenBank mRNA AK074258.1 - 7 - 7 1604 93
AK093986.1 - 1 - 1 1220 93
AL512717.1 - 2 - 2 648 93
AL832364.1 - 6 - 6 1516 93
AX775825.1 - 7 - 7 1604 93
BC015553.1 - 5 - 5 1008 93
BC027900.1 - 4 - 4 1100 93
BC062636.1 - 4 - 4 862 93
BC111056.1 - 7 - 7 1608 93

View Full Product Details