Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C16orf45
Entrez Gene ID
89927
Gene Name
chromosome 16 open reading frame 45
Gene Aliases
MINP
Chromosome Location
Chr.16: 15434468 - 15594521 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033201.2 NP_149978.1 1 - 2 296 64
XM_011522731.2 XP_011521033.1 1 - 2 275 64
XM_017023824.1 XP_016879313.1 1 - 2 273 64
GenBank mRNA AK057180.1 - 1 - 2 296 64
BC008967.2 - 1 - 2 226 64

View Full Product Details