Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYL10
Entrez Gene ID
93408
Gene Name
myosin light chain 10
Gene Aliases
MYLC2PL, PLRLC
Chromosome Location
Chr.7: 101613325 - 101629296 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138403.4 NP_612412.2 4 - 5 531 61
XM_017012793.1 XP_016868282.1 3 - 4 251 61
GenBank mRNA BC002778.2 - 4 - 5 531 61

View Full Product Details