Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEMT
Entrez Gene ID
10400
Gene Name
phosphatidylethanolamine N-methyltransferase
Gene Aliases
PEAMT, PEMPT, PEMT2, PNMT
Chromosome Location
Chr.17: 17505561 - 17591703 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267551.1 NP_001254480.1 5 - 6 583 124
NM_007169.2 NP_009100.2 5 - 6 641 124
NM_148172.2 NP_680477.1 5 - 6 649 124
NM_148173.1 NP_680478.1 5 - 6 528 124
XM_006721418.3 XP_006721481.2 5 - 6 530 124
XM_017024016.1 XP_016879505.1 3 - 4 436 124
GenBank mRNA AB029821.1 - 5 - 6 641 124
AF044214.1 - 5 - 6 614 124
AF113126.1 - 5 - 6 618 124
BC000557.2 - 5 - 6 625 124
BC050593.1 - 5 - 6 626 124
BM011377.1 - 5 - 6 511 124
CR457099.1 - 4 - 5 461 124

View Full Product Details