The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_148179.2 | NP_680545.1 | 1 - 2 | 252 | 69 |
GenBank mRNA | BQ224958.1 | - | 1 - 2 | 216 | 69 |
DT218980.1 | - | 1 - 2 | 220 | 69 |