The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152361.2 | NP_689574.1 | 1 - 1 | 67 | 73 |
GenBank mRNA | AK096263.1 | - | 1 - 1 | 54 | 73 |
BC009725.1 | - | 1 - 1 | 55 | 73 | |
DA382981.1 | - | 1 - 1 | 67 | 73 |