The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152367.2 | NP_689580.2 | 5 - 6 | 923 | 74 |
XM_011540623.2 | XP_011538925.1 | 5 - 6 | 1055 | 74 | |
XM_011540625.2 | XP_011538927.1 | 6 - 7 | 1151 | 74 | |
XM_011540626.2 | XP_011538928.1 | 5 - 6 | 977 | 74 | |
XM_017000260.1 | XP_016855749.1 | 6 - 7 | 2388 | 74 | |
GenBank mRNA | AK096035.1 | - | 5 - 6 | 74 | |
BC128148.1 | - | 4 - 5 | 74 | ||
BC128149.1 | - | 4 - 5 | 74 |