Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCA13
Entrez Gene ID
154664
Gene Name
ATP binding cassette subfamily A member 13
Gene Aliases
-
Chromosome Location
Chr.7: 48171424 - 48647497 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152701.4 NP_689914.3 55 - 56 14378 111
XM_011515130.2 XP_011513432.1 55 - 56 14413 111
XM_011515131.2 XP_011513433.1 55 - 56 14320 111
XM_011515132.2 XP_011513434.1 54 - 55 14320 111
XM_011515133.2 XP_011513435.1 54 - 55 14275 111
XM_011515134.2 XP_011513436.1 55 - 56 14413 111
XM_011515136.2 XP_011513438.2 50 - 51 13778 111
GenBank mRNA AF501281.1 - 37 - 38 5561 111
AK091270.1 - 3 - 4 381 111
AK128516.1 - 12 - 13 1673 111
AK131360.1 - 21 - 22 3150 111
AY204751.1 - 55 - 56 14379 111
JF913488.1 - 38 - 39 6276 111
JF913491.1 - 38 - 39 6183 111
JF913492.1 - 38 - 39 6276 111

View Full Product Details