Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPNE2
Entrez Gene ID
221184
Gene Name
copine 2
Gene Aliases
COPN2, CPN2
Chromosome Location
Chr.16: 57092543 - 57147966 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152727.5 NP_689940.3 2 - 3 462 77
GenBank mRNA AF492484.1 - 2 - 3 413 77
AK292269.1 - 1 - 2 371 77
AK315394.1 - 2 - 3 365 77
BC053658.1 - 3 - 4 536 77
BG830959.1 - 2 - 3 408 77
BG831872.1 - 2 - 3 410 77
CR749617.1 - 1 - 2 123 77

View Full Product Details