Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPNE2
Entrez Gene ID
221184
Gene Name
copine 2
Gene Aliases
COPN2, CPN2
Chromosome Location
Chr.16: 57092543 - 57147966 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152727.5 NP_689940.3 11 - 12 1345 62
GenBank mRNA AF492484.1 - 11 - 12 1296 62
AK094867.1 - 1 - 2 2689 62
AK126138.1 - 2 - 3 358 62
AK315394.1 - 11 - 12 1248 62
BC053658.1 - 12 - 13 1419 62
CR749617.1 - 10 - 11 1006 62

View Full Product Details