Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HIF3A
Entrez Gene ID
64344
Gene Name
hypoxia inducible factor 3 alpha subunit
Gene Aliases
HIF-3A, HIF3-alpha-1, IPAS, MOP7, PASD7, bHLHe17
Chromosome Location
Chr.19: 46297046 - 46343433 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152794.3 NP_690007.1 2 - 3 281 59
NM_152795.3 NP_690008.2 2 - 3 249 59
XM_005259152.4 XP_005259209.1 3 - 4 377 59
XM_005259155.4 XP_005259212.1 3 - 4 377 59
XM_005259156.4 XP_005259213.1 3 - 4 377 59
XM_017027132.1 XP_016882621.1 3 - 4 377 59
XM_017027133.1 XP_016882622.1 3 - 4 377 59
XM_017027134.1 XP_016882623.1 2 - 3 247 59
XM_017027135.1 XP_016882624.1 2 - 3 281 59
XM_017027136.1 XP_016882625.1 3 - 4 377 59
XM_017027137.1 XP_016882626.1 2 - 3 281 59
XM_017027138.1 XP_016882627.1 2 - 3 247 59
GenBank mRNA AB054067.1 - 2 - 3 212 59
AB118749.1 - 2 - 3 230 59
AB295039.1 - 2 - 3 230 59
AF079154.1 - 1 - 2 200 59
AF463492.1 - 2 - 3 244 59
AK021653.1 - 2 - 3 275 59
AK027725.1 - 2 - 3 230 59
AK304747.1 - 2 - 3 313 59
AK314680.1 - 2 - 3 281 59
AL541807.3 - 2 - 3 252 59
BC026308.1 - 2 - 3 240 59
BC080551.1 - 2 - 3 220 59
BI772109.1 - 2 - 3 306 59
DA562618.1 - 2 - 3 261 59
DB176660.1 - 2 - 3 249 59
DC423791.1 - 2 - 3 331 59
DR001068.1 - 2 - 3 257 59

View Full Product Details