The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022462.4 | NP_071907.4 | 1 - 2 | 138 | 78 |
NM_152796.2 | NP_690009.1 | 1 - 2 | 138 | 78 | |
XM_005259152.4 | XP_005259209.1 | 2 - 3 | 184 | 78 | |
XM_005259155.4 | XP_005259212.1 | 2 - 3 | 184 | 78 | |
XM_005259156.4 | XP_005259213.1 | 2 - 3 | 184 | 78 | |
XM_017027132.1 | XP_016882621.1 | 2 - 3 | 184 | 78 | |
XM_017027133.1 | XP_016882622.1 | 2 - 3 | 184 | 78 | |
XM_017027136.1 | XP_016882625.1 | 2 - 3 | 184 | 78 | |
XM_017027139.1 | XP_016882628.1 | 1 - 2 | 124 | 78 | |
XM_017027142.1 | XP_016882631.1 | 1 - 2 | 124 | 78 | |
GenBank mRNA | AK021421.1 | - | 1 - 2 | 88 | 78 |
AK021653.1 | - | 1 - 2 | 82 | 78 | |
AK024095.1 | - | 1 - 2 | 82 | 78 | |
AK297828.1 | - | 1 - 2 | 124 | 78 | |
AK304747.1 | - | 1 - 2 | 120 | 78 | |
BI772109.1 | - | 1 - 2 | 113 | 78 | |
DA559979.1 | - | 1 - 2 | 124 | 78 | |
DC423791.1 | - | 1 - 2 | 138 | 78 | |
DN989983.1 | - | 1 - 2 | 223 | 78 |