The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289000.1 | NP_001275929.1 | 19 - 19 | 3086 | 93 |
NM_001289001.1 | NP_001275930.1 | 18 - 18 | 3030 | 93 | |
NM_153609.3 | NP_705837.1 | 18 - 18 | 3099 | 93 | |
XM_011529989.2 | XP_011528291.1 | 16 - 16 | 2792 | 93 | |
GenBank mRNA | AW295507.1 | - | 1 - 1 | 93 | |
AY055384.1 | - | 18 - 18 | 3099 | 93 | |
AY358398.1 | - | 18 - 18 | 3030 | 93 |